The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128165.1 | NP_001121637.1 | 6 - 7 | 1077 | 177 |
NM_153214.2 | NP_694946.2 | 7 - 8 | 1215 | 177 | |
XM_011510585.2 | XP_011508887.1 | 7 - 8 | 1215 | 177 | |
XM_011510587.2 | XP_011508889.1 | 8 - 9 | 1192 | 177 | |
GenBank mRNA | AK094759.1 | - | 7 - 8 | 1215 | 177 |
AK124067.1 | - | 5 - 6 | 612 | 177 | |
BC035784.1 | - | 7 - 8 | 1168 | 177 | |
BC038242.1 | - | 3 - 4 | 410 | 177 | |
BC126986.1 | - | 6 - 7 | 879 | 177 | |
BC126987.1 | - | 4 - 5 | 615 | 177 |