Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO41
Entrez Gene ID
150726
Gene Name
F-box protein 41
Gene Aliases
FBX41
Chromosome Location
Chr.2: 73254682 - 73284453 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080410.2 NP_001073879.2 1 - 2 901 87
XM_006711950.2 XP_006712013.1 6 - 7 1885 87
XM_011532562.1 XP_011530864.1 2 - 3 1333 87
XM_017003421.1 XP_016858910.1 3 - 4 3982 87
XM_017003422.1 XP_016858911.1 6 - 7 3627 87
XM_017003423.1 XP_016858912.1 6 - 7 3008 87
GenBank mRNA AB075820.1 - 1 - 2 739 87

View Full Product Details