The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183387.2 | NP_899243.1 | 34 - 35 | 4782 | 150 |
XM_006720070.3 | XP_006720133.1 | 32 - 33 | 4558 | 150 | |
XM_011536528.2 | XP_011534830.1 | 34 - 35 | 4583 | 150 | |
XM_011536530.2 | XP_011534832.1 | 33 - 34 | 4573 | 150 | |
XM_011536531.2 | XP_011534833.1 | 32 - 33 | 4558 | 150 | |
XM_011536532.2 | XP_011534834.1 | 34 - 35 | 4582 | 150 | |
XM_011536533.2 | XP_011534835.1 | 32 - 33 | 4444 | 150 | |
XM_011536534.2 | XP_011534836.1 | 35 - 36 | 4500 | 150 | |
XM_011536536.2 | XP_011534838.1 | 34 - 35 | 4582 | 150 | |
XM_017021062.1 | XP_016876551.1 | 34 - 35 | 4582 | 150 | |
XM_017021063.1 | XP_016876552.1 | 33 - 34 | 4561 | 150 | |
XM_017021064.1 | XP_016876553.1 | 34 - 35 | 4468 | 150 | |
XM_017021065.1 | XP_016876554.1 | 31 - 32 | 4537 | 150 | |
XM_017021066.1 | XP_016876555.1 | 33 - 34 | 4383 | 150 | |
XM_017021067.1 | XP_016876556.1 | 34 - 35 | 4903 | 150 | |
XM_017021069.1 | XP_016876558.1 | 34 - 35 | 4827 | 150 | |
GenBank mRNA | AK127024.1 | - | 11 - 12 | 1252 | 150 |
AK128086.1 | - | 25 - 26 | 3297 | 150 | |
BC150639.1 | - | 34 - 35 | 4782 | 150 |