Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP8A1
Entrez Gene ID
10396
Gene Name
ATPase phospholipid transporting 8A1
Gene Aliases
ATPASEII, ATPIA, ATPP2
Chromosome Location
Chr.4: 42408373 - 42657134 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105529.1 NP_001098999.1 17 - 18 1792 139
NM_006095.2 NP_006086.1 18 - 19 1837 139
XM_005248043.2 XP_005248100.1 18 - 19 1832 139
XM_011513615.1 XP_011511917.1 18 - 19 1832 139
XM_011513616.2 XP_011511918.1 18 - 19 1866 139
XM_011513618.1 XP_011511920.1 17 - 18 1787 139
XM_017007645.1 XP_016863134.1 17 - 18 1821 139
XM_017007646.1 XP_016863135.1 17 - 18 1739 139
XM_017007647.1 XP_016863136.1 16 - 17 1692 139
GenBank mRNA AB013452.1 - 18 - 19 1618 139
AB209687.1 - 17 - 18 1645 139
AF067820.1 - 18 - 19 1605 139
BC109317.1 - 17 - 18 1554 139
BC109318.1 - 17 - 18 1560 139

View Full Product Details