The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105529.1 | NP_001098999.1 | 30 - 31 | 3149 | 84 |
NM_006095.2 | NP_006086.1 | 31 - 32 | 3194 | 84 | |
XM_005248043.2 | XP_005248100.1 | 31 - 32 | 3189 | 84 | |
XM_011513615.1 | XP_011511917.1 | 32 - 33 | 3210 | 84 | |
XM_011513616.2 | XP_011511918.1 | 32 - 33 | 3244 | 84 | |
XM_011513618.1 | XP_011511920.1 | 31 - 32 | 3165 | 84 | |
XM_017007645.1 | XP_016863134.1 | 30 - 31 | 3178 | 84 | |
XM_017007646.1 | XP_016863135.1 | 31 - 32 | 3117 | 84 | |
XM_017007647.1 | XP_016863136.1 | 29 - 30 | 3049 | 84 | |
GenBank mRNA | AB013452.1 | - | 31 - 32 | 2975 | 84 |
AB209687.1 | - | 30 - 31 | 3002 | 84 | |
AF067820.1 | - | 31 - 32 | 2962 | 84 | |
BC109317.1 | - | 30 - 31 | 2911 | 84 | |
BC109318.1 | - | 30 - 31 | 2917 | 84 | |
BX648675.1 | - | 8 - 9 | 842 | 84 |