The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113402.1 | NP_001106873.1 | 2 - 3 | 338 | 91 |
NM_001278411.1 | NP_001265340.1 | 2 - 3 | 318 | 91 | |
NM_001278412.1 | NP_001265341.1 | 4 - 5 | 547 | 91 | |
XM_017018963.1 | XP_016874452.1 | 4 - 5 | 387 | 91 | |
XM_017018964.1 | XP_016874453.1 | 2 - 3 | 320 | 91 | |
XM_017018965.1 | XP_016874454.1 | 3 - 4 | 412 | 91 | |
GenBank mRNA | AK123033.1 | - | 2 - 3 | 185 | 91 |
AK302112.1 | - | 4 - 5 | 394 | 91 | |
AL833434.1 | - | 2 - 3 | 188 | 91 | |
BC037897.2 | - | 2 - 3 | 242 | 91 | |
BC045831.1 | - | 2 - 3 | 282 | 91 | |
BC067906.1 | - | 2 - 3 | 193 | 91 | |
BI464068.1 | - | 2 - 3 | 267 | 91 | |
DC325581.1 | - | 2 - 3 | 244 | 91 | |
DC394802.1 | - | 2 - 3 | 338 | 91 |