The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011547.2 | NP_001011547.2 | 2 - 3 | 283 | 140 |
NM_001135181.1 | NP_001128653.1 | 2 - 3 | 283 | 140 | |
XM_011540924.2 | XP_011539226.1 | 2 - 3 | 1850 | 140 | |
XM_011540925.2 | XP_011539227.1 | 2 - 3 | 1851 | 140 | |
XM_011540926.2 | XP_011539228.1 | 2 - 3 | 271 | 140 | |
XM_011540928.1 | XP_011539230.1 | 2 - 3 | 229 | 140 | |
XM_011540929.2 | XP_011539231.1 | 2 - 3 | 1855 | 140 | |
XM_017000558.1 | XP_016856047.1 | 2 - 3 | 1854 | 140 | |
GenBank mRNA | AK131200.1 | - | 2 - 3 | 236 | 140 |
AK296133.1 | - | 2 - 3 | 229 | 140 | |
BC104857.1 | - | 2 - 3 | 283 | 140 | |
BC104863.1 | - | 2 - 3 | 283 | 140 | |
BC143570.1 | - | 2 - 3 | 283 | 140 | |
BC143572.1 | - | 2 - 3 | 281 | 140 | |
BC143587.1 | - | 2 - 3 | 283 | 140 |