The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011547.2 | NP_001011547.2 | 3 - 4 | 392 | 151 |
XM_011540928.1 | XP_011539230.1 | 3 - 4 | 338 | 151 | |
GenBank mRNA | AK131200.1 | - | 3 - 4 | 345 | 151 |
AK296133.1 | - | 3 - 4 | 338 | 151 | |
BC104857.1 | - | 3 - 4 | 392 | 151 | |
BC104863.1 | - | 3 - 4 | 392 | 151 | |
BC143587.1 | - | 3 - 4 | 392 | 151 |