The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256169.1 | NP_001243098.1 | 3 - 4 | 372 | 84 |
NM_019101.2 | NP_061974.2 | 3 - 4 | 417 | 84 | |
NR_045828.1 | 3 - 4 | 379 | 84 | ||
XM_006715150.3 | XP_006715213.1 | 3 - 4 | 401 | 84 | |
GenBank mRNA | AA846749.1 | - | 3 - 4 | 84 | |
AF118393.3 | - | 3 - 4 | 377 | 84 | |
AF161454.1 | - | 3 - 4 | 338 | 84 | |
AJ245434.1 | - | 3 - 4 | 393 | 84 | |
BC020683.1 | - | 3 - 4 | 396 | 84 | |
CB054016.1 | - | 3 - 4 | 379 | 84 | |
CN428415.1 | - | 3 - 4 | 359 | 84 |