The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004075.4 | NP_004066.1 | 3 - 4 | 1305 | 126 |
XM_017018832.1 | XP_016874321.1 | 4 - 5 | 1440 | 126 | |
GenBank mRNA | AK125915.1 | - | 5 - 6 | 968 | 126 |
AK290552.1 | - | 3 - 4 | 605 | 126 | |
BC017466.1 | - | 3 - 4 | 728 | 126 | |
BC030519.1 | - | 3 - 4 | 978 | 126 | |
D83702.1 | - | 3 - 4 | 995 | 126 | |
D84657.1 | - | 3 - 4 | 409 | 126 |