The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194449.3 | NP_919431.2 | 4 - 5 | 2316 | 148 |
XM_011525886.1 | XP_011524188.1 | 4 - 5 | 2310 | 148 | |
GenBank mRNA | AB011178.2 | - | 4 - 5 | 1029 | 148 |
BC014927.2 | - | 4 - 5 | 603 | 148 | |
BC126277.1 | - | 4 - 5 | 651 | 148 | |
BG110729.1 | - | - | 321 | 147 |