The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003850.2 | NP_003841.1 | 5 - 6 | 721 | 89 |
XM_017020806.1 | XP_016876295.1 | 5 - 6 | 941 | 89 | |
XM_017020807.1 | XP_016876296.1 | 7 - 8 | 956 | 89 | |
XM_017020808.1 | XP_016876297.1 | 5 - 6 | 708 | 89 | |
XM_017020809.1 | XP_016876298.1 | 6 - 7 | 805 | 89 | |
GenBank mRNA | AB035863.1 | - | 5 - 6 | 721 | 89 |
AF058953.1 | - | 4 - 5 | 553 | 89 | |
AF087890.1 | - | 5 - 6 | 729 | 89 | |
AK001458.1 | - | 5 - 6 | 619 | 89 | |
AK001847.1 | - | 5 - 6 | 708 | 89 | |
AK225452.1 | - | 5 - 6 | 708 | 89 | |
AK299437.1 | - | 5 - 6 | 660 | 89 | |
AK302311.1 | - | 6 - 7 | 780 | 89 | |
AK315513.1 | - | 5 - 6 | 683 | 89 | |
AK316041.1 | - | 6 - 7 | 635 | 89 | |
AK316271.1 | - | 6 - 7 | 1036 | 89 | |
BC027587.2 | - | 5 - 6 | 673 | 89 | |
BX640602.1 | - | 5 - 6 | 692 | 89 | |
KU178442.1 | - | 5 - 6 | 664 | 89 | |
KU178443.1 | - | 2 - 3 | 220 | 89 |