Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPP6R2
Entrez Gene ID
9701
Gene Name
protein phosphatase 6 regulatory subunit 2
Gene Aliases
KIAA0685, PP6R2, SAP190, SAPS2
Chromosome Location
Chr.22: 50343317 - 50445090 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242898.1 NP_001229827.1 16 - 17 2158 187
XM_005261960.3 XP_005262017.1 10 - 11 1112 187
XM_006724431.2 XP_006724494.1 17 - 18 2269 187
XM_006724434.1 XP_006724497.1 18 - 19 2418 187
XM_011530720.2 XP_011529022.1 15 - 16 1846 187
XM_011530721.2 XP_011529023.1 15 - 16 1843 187
XM_011530722.2 XP_011529024.1 15 - 16 1843 187
XM_011530723.2 XP_011529025.1 15 - 16 1846 187
XM_011530724.2 XP_011529026.1 15 - 16 1846 187
XM_011530732.2 XP_011529034.1 18 - 19 2322 187
XM_011530734.2 XP_011529036.1 17 - 18 2196 187
XM_011530735.2 XP_011529037.1 16 - 17 2146 187
XM_011530736.2 XP_011529038.1 17 - 18 2272 187
XM_011530737.2 XP_011529039.1 18 - 19 2421 187
XM_011530738.2 XP_011529040.1 17 - 18 2224 187
XM_011530739.2 XP_011529041.1 15 - 16 1886 187
XM_011530740.2 XP_011529042.1 17 - 18 2294 187
XM_017029117.1 XP_016884606.1 19 - 20 2471 187
XM_017029118.1 XP_016884607.1 19 - 20 2423 187
XM_017029119.1 XP_016884608.1 18 - 19 2345 187
XM_017029120.1 XP_016884609.1 18 - 19 2319 187
XM_017029121.1 XP_016884610.1 16 - 17 2143 187
XM_017029122.1 XP_016884611.1 17 - 18 2221 187
XM_017029123.1 XP_016884612.1 16 - 17 2143 187
XM_017029124.1 XP_016884613.1 17 - 18 2269 187
XM_017029125.1 XP_016884614.1 16 - 17 2143 187
XM_017029126.1 XP_016884615.1 16 - 17 2146 187
XM_017029127.1 XP_016884616.1 17 - 18 2221 187
XM_017029128.1 XP_016884617.1 17 - 18 2269 187
XM_017029133.1 XP_016884622.1 16 - 17 2100 187
XM_017029134.1 XP_016884623.1 16 - 17 2097 187
XM_017029135.1 XP_016884624.1 17 - 18 2147 187
XM_017029136.1 XP_016884625.1 16 - 17 2097 187
XM_017029137.1 XP_016884626.1 16 - 17 2097 187
GenBank mRNA AK002063.1 - 9 - 10 954 187
AK302472.1 - 16 - 17 2069 187
BC000976.4 - 17 - 18 2147 187

View Full Product Details