Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD99P1
Entrez Gene ID
401577
Gene Name
CD99 molecule pseudogene 1
Gene Aliases
CD99L1, CXYorf12, MIC2R, NCRNA00103
Chromosome Location
Chr.Y: 2609265 - 2657229 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033381.1 7 - 8 644 149
GenBank mRNA AK023533.1 - 3 - 4 2282 149
BC019893.1 - 7 - 8 515 149

View Full Product Details