The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_170733.3 | NP_733929.1 | 1 - 1 | 170 | 147 |
GenBank mRNA | AB038672.1 | - | 1 - 1 | 52 | 147 |
AY054397.2 | - | 1 - 1 | 52 | 147 | |
BC029795.1 | - | 1 - 1 | 143 | 147 | |
EF674521.1 | - | 1 - 1 | 170 | 147 |