The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012367.1 | NP_036499.1 | 1 - 1 | 448 | 88 |
GenBank mRNA | BC109251.1 | - | 1 - 1 | 448 | 88 |
BC110457.2 | - | 1 - 1 | 448 | 88 |