The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172089.3 | NP_742086.1 | 6 - 7 | 589 | 86 |
GenBank mRNA | AY081051.1 | - | 6 - 7 | 548 | 86 |