Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP4F22
Entrez Gene ID
126410
Gene Name
cytochrome P450 family 4 subfamily F member 22
Gene Aliases
ARCI5, INLNE, LI3
Chromosome Location
Chr.19: 15508487 - 15552317 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173483.3 NP_775754.2 4 - 5 530 83
XM_011527692.2 XP_011525994.1 5 - 6 772 83
XM_011527693.2 XP_011525995.1 4 - 5 542 83
GenBank mRNA AK096820.1 - 4 - 5 83
BC069351.1 - 3 - 4 467 83
BC093894.1 - 3 - 4 404 83
BC093896.1 - 3 - 4 404 83

View Full Product Details