Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LNPEP
Entrez Gene ID
4012
Gene Name
leucyl and cystinyl aminopeptidase
Gene Aliases
CAP, IRAP, P-LAP, PLAP
Chromosome Location
Chr.5: 96935642 - 97029411 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK096804.1 - 1 - 2 303 109

View Full Product Details