Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASB16-AS1
Entrez Gene ID
339201
Gene Name
ASB16 antisense RNA 1
Gene Aliases
C17orf65
Chromosome Location
Chr.17: 44175973 - 44186717 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_049729.1 4 - 4 1815 71
NR_049730.1 3 - 3 477 71
GenBank mRNA AK055685.1 - 4 - 4 1769 71
BC009448.1 - 1 - 1 897 71

View Full Product Details