The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039372.2 | NP_001034461.1 | 7 - 8 | 1274 | 66 |
NM_001288804.1 | NP_001275733.1 | 8 - 9 | 1343 | 66 | |
NM_001288810.1 | NP_001275739.1 | 5 - 6 | 1241 | 66 | |
NM_198151.2 | NP_937794.1 | 6 - 7 | 1266 | 66 | |
XM_011516000.2 | XP_011514302.1 | 7 - 8 | 1207 | 66 | |
XM_011516001.1 | XP_011514303.1 | 7 - 8 | 1207 | 66 | |
GenBank mRNA | AK096002.1 | - | 7 - 8 | 1227 | 66 |
AK298361.1 | - | 8 - 9 | 1293 | 66 | |
AY358345.1 | - | 6 - 7 | 1228 | 66 | |
BC136300.1 | - | 6 - 7 | 1196 | 66 | |
BC136301.1 | - | 6 - 7 | 1196 | 66 | |
BC139906.1 | - | 6 - 7 | 1253 | 66 |