Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EYS
Entrez Gene ID
346007
Gene Name
eyes shut homolog (Drosophila)
Gene Aliases
C6orf178, C6orf179, C6orf180, EGFL10, EGFL11, RP25, SPAM, bA166P24.2, bA307F22.3, bA74E24.1, dJ1018A4.2, dJ22I17.2, dJ303F19.1
Chromosome Location
Chr.6: 63719980 - 65707225 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142800.1 NP_001136272.1 7 - 8 1721 131
NM_001142801.1 NP_001136273.1 7 - 8 1721 131
NM_001292009.1 NP_001278938.1 7 - 8 1721 131
NM_198283.1 NP_938024.1 6 - 7 1580 131
GenBank mRNA AB593114.1 - 7 - 8 1686 131
AY358133.1 - 6 - 7 1580 131
BC133011.1 - 5 - 6 1445 131
BC133013.1 - 5 - 6 1442 131
BX537914.1 - 7 - 8 1721 131
FJ416331.1 - 7 - 8 1729 131
FM209056.1 - 7 - 8 1673 131

View Full Product Details