Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EYS
Entrez Gene ID
346007
Gene Name
eyes shut homolog (Drosophila)
Gene Aliases
C6orf178, C6orf179, C6orf180, EGFL10, EGFL11, RP25, SPAM, bA166P24.2, bA307F22.3, bA74E24.1, dJ1018A4.2, dJ22I17.2, dJ303F19.1
Chromosome Location
Chr.6: 63719980 - 65707225 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142800.1 NP_001136272.1 10 - 11 2140 72
NM_001142801.1 NP_001136273.1 10 - 11 2140 72
NM_001292009.1 NP_001278938.1 10 - 11 2140 72
NM_198283.1 NP_938024.1 9 - 10 1999 72
GenBank mRNA AB593114.1 - 10 - 11 2105 72
AY358133.1 - 9 - 10 1999 72
BC133011.1 - 8 - 9 1864 72
BC133013.1 - 8 - 9 1861 72
FJ416331.1 - 10 - 11 2148 72
FM209056.1 - 10 - 11 2092 72

View Full Product Details