Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RUFY4
Entrez Gene ID
285180
Gene Name
RUN and FYVE domain containing 4
Gene Aliases
ZFYVE31
Chromosome Location
Chr.2: 218034934 - 218090584 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198483.3 NP_940885.2 9 - 10 1765 95
NR_034176.1 10 - 11 2626 95
XM_011511014.2 XP_011509316.1 11 - 12 2707 95
XM_017003895.1 XP_016859384.1 10 - 11 2842 95
XM_017003896.1 XP_016859385.1 10 - 11 2839 95
XM_017003897.1 XP_016859386.1 10 - 11 2842 95
GenBank mRNA AK128393.1 - 10 - 11 2572 95
AK131246.1 - 8 - 9 1717 95
BC113712.1 - 3 - 4 787 95

View Full Product Details