Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RUFY4
Entrez Gene ID
285180
Gene Name
RUN and FYVE domain containing 4
Gene Aliases
ZFYVE31
Chromosome Location
Chr.2: 218034934 - 218090584 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_034176.1 7 - 8 1764 127
GenBank mRNA AK128393.1 - 7 - 8 1710 127

View Full Product Details