Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFAP54
Entrez Gene ID
144535
Gene Name
cilia and flagella associated protein 54
Gene Aliases
C12orf55, C12orf63
Chromosome Location
Chr.12: 96489566 - 96875555 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306084.1 NP_001293013.1 52 - 53 7253 66
XM_011539069.2 XP_011537371.1 53 - 54 7453 66
XM_011539070.2 XP_011537372.1 53 - 54 7453 66
XM_011539071.2 XP_011537373.1 53 - 54 7453 66
XM_011539072.2 XP_011537374.1 53 - 54 7453 66
XM_011539073.2 XP_011537375.1 53 - 54 7453 66
XM_011539074.2 XP_011537376.1 53 - 54 7453 66
XM_011539076.2 XP_011537378.1 53 - 54 7453 66
XM_011539077.2 XP_011537379.1 52 - 53 7603 66
XM_011539078.2 XP_011537380.1 53 - 54 7453 66
XM_011539079.1 XP_011537381.1 49 - 50 6699 66
XM_011539081.2 XP_011537383.1 53 - 54 7453 66
XM_011539083.2 XP_011537385.1 53 - 54 7453 66
XM_011539085.1 XP_011537387.1 38 - 39 5274 66
XM_017018866.1 XP_016874355.1 53 - 54 7453 66
XM_017018867.1 XP_016874356.1 42 - 43 5705 66
GenBank mRNA AK126100.1 - 19 - 20 2567 66
DB225896.1 - 3 - 4 421 66

View Full Product Details