The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017975.4 | NP_001017975.4 | 28 - 29 | 3236 | 106 |
XM_011540849.1 | XP_011539151.1 | 28 - 29 | 3319 | 106 | |
XM_011540850.2 | XP_011539152.1 | 28 - 29 | 3248 | 106 | |
XM_011540851.1 | XP_011539153.1 | 28 - 29 | 3203 | 106 | |
XM_011540852.2 | XP_011539154.1 | 28 - 29 | 3185 | 106 | |
XM_011540855.1 | XP_011539157.1 | 26 - 27 | 3041 | 106 | |
XM_011540857.1 | XP_011539159.1 | 26 - 27 | 2800 | 106 | |
XM_011540859.2 | XP_011539161.1 | 25 - 26 | 2769 | 106 | |
XM_017000490.1 | XP_016855979.1 | 27 - 28 | 3099 | 106 | |
XM_017000491.1 | XP_016855980.1 | 27 - 28 | 3048 | 106 | |
XM_017000492.1 | XP_016855981.1 | 22 - 23 | 2283 | 106 | |
XM_017000493.1 | XP_016855982.1 | 16 - 17 | 1487 | 106 | |
GenBank mRNA | AB044787.1 | - | 5 - 6 | 524 | 106 |
AB204867.1 | - | 28 - 29 | 3237 | 106 | |
AK094079.1 | - | 11 - 12 | 1063 | 106 | |
AK294754.1 | - | 22 - 23 | 2253 | 106 | |
AK302582.1 | - | 24 - 25 | 2404 | 106 | |
BC130525.1 | - | 13 - 14 | 1182 | 106 | |
BC144172.1 | - | 13 - 14 | 1182 | 106 | |
BX649037.1 | - | 9 - 10 | 772 | 106 |