The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_201613.2 | NP_963907.1 | 1 - 2 | 532 | 132 |
GenBank mRNA | AJ539427.1 | - | 1 - 2 | 292 | 132 |
AK123069.1 | - | 1 - 2 | 436 | 132 | |
BC029415.1 | - | 1 - 2 | 292 | 132 | |
BC051372.1 | - | 1 - 2 | 261 | 132 |