The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003969.3 | NP_003960.1 | 1 - 1 | 262 | 82 |
GenBank mRNA | BM559596.1 | - | - | 254 | 82 |
BU508830.1 | - | 1 - 1 | 206 | 82 |