Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HNRNPA1
Entrez Gene ID
3178
Gene Name
heterogeneous nuclear ribonucleoprotein A1
Gene Aliases
ALS19, ALS20, HNRPA1, HNRPA1L3, IBMPFD3, UP 1, hnRNP A1, hnRNP-A1
Chromosome Location
Chr.12: 54280690 - 54287088 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002136.3 NP_002127.1 1 - 1 68 81
NM_031157.3 NP_112420.1 1 - 1 68 81
NR_135167.1 1 - 1 68 81
XM_005268826.1 XP_005268883.1 1 - 1 62 81
XM_017019251.1 XP_016874740.1 1 - 1 62 81
GenBank mRNA BQ277208.1 - - 68 81
CD705336.1 - 1 - 1 85 81

View Full Product Details