Product Details

Assay Reference Genome
Location

Chr.22:38113649 on build GRCh38
Cytoband
22q13.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
8398
Gene Symbol
PLA2G6
Gene Name
phospholipase A2 group VI
Gene Aliases
CaI-PLA2, GVI, INAD1, IPLA2-VIA, NBIA2, NBIA2A, NBIA2B, PARK14, PLA2, PNPLA9, iPLA2, iPLA2beta
Location
Chr.22:38111495-38192099 on build GRCh38
Assay Gene Location
Overlaps Intron 22 - Exon 23
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
PLA2G6 NM_001004426.1 NP_001004426.1
NM_001199562.1 NP_001186491.1
NM_003560.2 NP_003551.2
XM_005261764.2 XP_005261821.1
XM_005261765.1 XP_005261822.1
XM_005261766.1 XP_005261823.1
XM_006724332.3 XP_006724395.1
XM_011530422.1 XP_011528724.1
XM_011530423.1 XP_011528725.1
XM_011530424.1 XP_011528726.1
XM_011530425.1 XP_011528727.1
XM_017028981.1 XP_016884470.1
XM_017028982.1 XP_016884471.1
XM_017028983.1 XP_016884472.1
XM_017028984.1 XP_016884473.1
XM_017028985.1 XP_016884474.1
AF064594.1 AAC97486.1
AF102988.1 AAD41722.1
AF102989.1 AAD41723.1
AK096521.1
AK128862.1
AK290825.1
AK291212.1
AK308308.1
AL080187.1 CAB45768.2
BC036742.1 AAH36742.2
BC051904.1 AAH51904.1
CR456543.1 CAG30429.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv471198 Chr22:38019714 - 38149612 on Build GRCh38 Loss PLA2G6 BAIAP2L2 SLC16A8 PICK1 POLR2F
nsv3623 Chr22:38105535 - 38113801 on Build GRCh38 Insertion PLA2G6 BAIAP2L2
nsv508732 Chr22:38096355 - 38180191 on Build GRCh38 Insertion PLA2G6 BAIAP2L2
nsv834194 Chr22:38063495 - 38240504 on Build GRCh38 Loss MAFF PLA2G6 SNORA92 TMEM184B BAIAP2L2 SLC16A8 PICK1
nsv834193 Chr22:37997329 - 38172090 on Build GRCh38 Loss PLA2G6 BAIAP2L2 SLC16A8 PICK1 POLR2F

View Full Product Details