The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_033415.1 | 9 - 10 | 2510 | 61 | |
GenBank mRNA | AK125965.1 | - | 9 - 10 | 2510 | 61 |