Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HECW1-IT1
Entrez Gene ID
100127950
Gene Name
HECW1 intronic transcript 1
Gene Aliases
-
Chromosome Location
Chr.7: 43117896 - 43163187 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_135295.1 2 - 3 927 69
GenBank mRNA AK093262.1 - 2 - 3 927 69

View Full Product Details