Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR51B6
Entrez Gene ID
390058
Gene Name
olfactory receptor family 51 subfamily B member 6
Gene Aliases
HOR5'Beta6
Chromosome Location
Chr.11: 5351508 - 5352446 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004750.1 NP_001004750.1 1 - 1 896 99

View Full Product Details