The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005567.2 | NP_001005567.2 | 3 - 4 | 214 | 130 |
NR_038321.1 | 3 - 4 | 210 | 130 | ||
GenBank mRNA | BC022184.1 | - | 3 - 4 | 214 | 130 |
BG493214.1 | - | 3 - 4 | 210 | 130 | |
BG776801.1 | - | 3 - 4 | 210 | 130 |