The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000474.3 | NP_000465.1 | 1 - 1 | 953 | 85 |
XM_011515496.1 | XP_011513798.1 | 1 - 1 | 700 | 85 | |
GenBank mRNA | BC036704.2 | - | 1 - 1 | 953 | 85 |
X99268.1 | - | 1 - 1 | 84 |