Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBE2MP1
Entrez Gene ID
606551
Gene Name
ubiquitin conjugating enzyme E2 M pseudogene 1
Gene Aliases
-
Chromosome Location
Chr.16: 35169431 - 35170391 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_002837.1 1 - 1 619 94

View Full Product Details