Product Details

Assay Reference Genome
Location

Chr.7:101031762 on build GRCh38
Cytoband
7q22.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
140453
Gene Symbol
MUC17
Gene Name
mucin 17, cell surface associated
Gene Aliases
MUC-17, MUC-3, MUC3
Location
Chr.7:101020083-101058859 on build GRCh38
Assay Gene Location
Within Exon 3
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
MUC17 NM_001040105.1 3 399 NP_001035194.1
NR_133665.1 3 399
AJ606307.1 3 399 CAE54435.1
AJ606308.1 3 399 CAE54436.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv428181 Chr7:100962999 - 101105965 on Build GRCh38 Gain+Loss MUC12 LOC102724094 MUC17 MUC3A TRIM56
esv2759547 Chr7:100757867 - 101105965 on Build GRCh38 Gain+Loss TRIP6 UFSP1 SLC12A9 TRIM56 SRRT MUC12 LOC102724094 MIR6875 ACHE MUC17 MUC3A EPHB4 ZAN
esv2734888 Chr7:99975623 - 101290285 on Build GRCh38 Deletion STAG3L5P ACTL6B CLDN15 AGFG2 LAMTOR4 C7orf43 TSC22D4 VGF STAG3 GPC2 MUC12 PVRIG LOC102724094 MBLAC1 PMS2P1 ACHE MCM7 SPDYE3 NAT16 ZSCAN21 AP1S1 ZKSCAN1 ZNF3 SERPINE1 MOGAT3 MOSPD3 SAP25 GAL3ST4 PCOLCE-AS1 PLOD3 MUC3A ZAN GNB2 GIGYF1 TRIP6 AZGP1 MIR93 UFSP1 SLC12A9 AP4M1 FIS1 LRCH4 MIR4658 ZASP ZCWPW1 PCOLCE MIR6875 ZNHIT1 TAF6 EPHB4 MIR6840 PPP1R35 MIR106B MIR4653 MEPCE FBXO24 PILRB AZGP1P1 LOC105375429 PILRA COPS6 TRIM56 MIR25 SRRT C7orf61 EPO GATS PVRIG2P CNPY4 NYAP1 POP7 STAG3L5P-PVRIG2P-PILRB MUC17 TFR2
nsv820525 Chr7:101030087 - 101043704 on Build GRCh38 Duplication MUC17
nsv1033081 Chr7:101026348 - 101128543 on Build GRCh38 Gain MUC17 TRIM56 SERPINE1
nsv831074 Chr7:100963006 - 101102251 on Build GRCh38 Loss MUC12 LOC102724094 MUC17 MUC3A TRIM56

View Full Product Details