Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIPSL
Entrez Gene ID
266971
Gene Name
PIP5K1A and PSMD4-like, pseudogene
Gene Aliases
PIP5K1L1, PIP5K1P3, PSMD4P2, bA429H9.1
Chromosome Location
Chr.10: 93958140 - 93961915 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_002319.2 1 - 1 2046 113

View Full Product Details