The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001141968.1 | NP_001135440.1 | 17 - 18 | 1466 | 147 |
NM_001271850.1 | NP_001258779.1 | 5 - 6 | 745 | 147 | |
NM_130785.3 | NP_570141.3 | 16 - 17 | 1376 | 147 | |
NM_199254.2 | NP_954863.2 | 19 - 20 | 1607 | 147 | |
NR_073487.1 | 13 - 14 | 1144 | 147 | ||
GenBank mRNA | AJ421032.1 | - | 16 - 17 | 1376 | 147 |
AY219890.1 | - | 19 - 20 | 1607 | 147 | |
BC128146.1 | - | 17 - 18 | 1448 | 147 | |
BC128147.1 | - | 13 - 14 | 1126 | 147 | |
FJ969729.1 | - | 5 - 6 | 748 | 147 |