Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FAM66A
- Entrez Gene ID
-
100133172
- Gene Name
- family with sequence similarity 66 member A
- Gene Aliases
-
-
- Chromosome Location
-
Chr.8: 12362019 - 12411001 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_026789.1 |
|
1 - 1 |
265 |
117 |
GenBank mRNA |
AK311040.1 |
- |
1 - 1 |
266 |
117 |
DC372067.1 |
- |
1 - 1 |
|
116 |
- Gene Symbol
- FAM66D
- Entrez Gene ID
-
100132923
- Gene Name
- family with sequence similarity 66 member D
- Gene Aliases
-
-
- Chromosome Location
-
Chr.8: 12115782 - 12151189 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027425.1 |
|
1 - 1 |
265 |
117 |
GenBank mRNA |
AK294595.1 |
- |
1 - 1 |
265 |
117 |
View Full Product Details