Product Details

Assay Reference Genome
Location

Chr.8:38413222 on build GRCh38
Cytoband
8p11.23
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
2260
Gene Symbol
FGFR1
Gene Name
fibroblast growth factor receptor 1
Gene Aliases
BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1
Location
Chr.8:38411138-38468834 on build GRCh38
Assay Gene Location
Within Exon 23
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
FGFR1 NM_001174063.1 18 3811 NP_001167534.1
NM_001174064.1 19 3879 NP_001167535.1
NM_001174065.1 18 3131 NP_001167536.1
NM_001174066.1 17 2870 NP_001167537.1
NM_001174067.1 19 3291 NP_001167538.1
NM_015850.3 18 3811 NP_056934.2
NM_023105.2 17 3550 NP_075593.1
NM_023106.2 17 3544 NP_075594.1
NM_023110.2 18 3817 NP_075598.2
XM_006716303.2 XP_006716366.1
XM_006716304.1 XP_006716367.1
XM_006716306.2 XP_006716369.1
XM_006716307.1 XP_006716370.1
XM_006716309.3 XP_006716372.1
XM_006716310.2 XP_006716373.1
XM_006716311.1 XP_006716374.1
XM_006716312.1 XP_006716375.1
XM_006716313.2 XP_006716376.1
XM_006716314.1 XP_006716377.1
XM_011544443.2 XP_011542745.1
XM_011544444.1 XP_011542746.1
XM_011544445.2 XP_011542747.1
XM_011544446.2 18 3118 XP_011542748.1
XM_011544447.2 18 3112 XP_011542749.1
XM_011544448.1 XP_011542750.1
XM_011544449.1 XP_011542751.1
XM_011544450.2 17 2716 XP_011542752.1
XM_011544451.1 XP_011542753.1
XM_017013219.1 XP_016868708.1
XM_017013220.1 18 3106 XP_016868709.1
XM_017013221.1 XP_016868710.1
XM_017013222.1 XP_016868711.1
XM_017013223.1 XP_016868712.1
XM_017013224.1 18 3606 XP_016868713.1
XM_017013225.1 18 3089 XP_016868714.1
XM_017013226.1 17 2716 XP_016868715.1
XM_017013227.1 17 2710 XP_016868716.1
XM_017013228.1 17 3339 XP_016868717.1
XM_017013229.1 XP_016868718.1
XM_017013230.1 16 3904 XP_016868719.1
AB208919.1 19 3666 BAD92156.1
AK222718.1 18 3610 BAD96438.1
AK292470.1
BC015035.1 19 3539 AAH15035.1
BC018128.1 18 3575 AAH18128.1
BC091494.1 16 2681 AAH91494.1
BX649167.1 14 5159
FJ809917.1 18 3826
M34185.1 17 2863 AAA35836.1
M34186.1 17 3128 AAA35837.1
M34641.1 17 2878 AAA35835.1
M37722.1 17 2830 AAA75007.1
M60485.1 19 3540 AAA35840.1
M63887.1 17 2875 AAA35958.1
M63888.1 17 2900 AAA35959.1
X66945.1 18 3601 CAA47375.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv951128 Chr8:38411783 - 38418282 on Build GRCh38 Deletion FGFR1

View Full Product Details