The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308437.1 | NP_001295366.1 | 4 - 4 | 1088 | 119 |
NM_173635.2 | NP_775906.1 | 6 - 6 | 1561 | 119 | |
NR_102324.1 | 5 - 5 | 1565 | 119 | ||
XM_005258799.4 | XP_005258856.1 | 6 - 6 | 1852 | 119 | |
XM_011526821.2 | XP_011525123.1 | 6 - 6 | 1474 | 119 | |
XM_017026647.1 | XP_016882136.1 | 5 - 5 | 1567 | 119 | |
GenBank mRNA | AK097554.1 | - | 6 - 6 | 1561 | 119 |
DB515179.2 | - | 1 - 1 | 540 | 119 |