The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004983.2 | NP_004974.2 | 3 - 3 | 2981 | 67 |
XM_017001247.1 | XP_016856736.1 | 8 - 8 | 9663 | 67 | |
GenBank mRNA | U52152.1 | - | 3 - 3 | 2997 | 67 |