The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_028336.1 | 2 - 2 | 250 | 111 | |
GenBank mRNA | BC031617.1 | - | 2 - 2 | 250 | 111 |