The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005130.4 | NP_005121.1 | 3 - 3 | 1108 | 143 |
GenBank mRNA | AK291307.1 | - | 2 - 2 | 930 | 143 |
BC003628.2 | - | 2 - 2 | 910 | 143 | |
BC008910.2 | - | 2 - 2 | 921 | 143 | |
BC018852.2 | - | 2 - 2 | 922 | 143 | |
BX434467.2 | - | 1 - 1 | 143 | ||
M60047.1 | - | 2 - 2 | 910 | 143 |