Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MT3
Entrez Gene ID
4504
Gene Name
metallothionein 3
Gene Aliases
GIF, GIFB, GRIF, ZnMT3
Chromosome Location
Chr.16: 56589355 - 56591088 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005954.2 NP_005945.1 3 - 3 541 81
GenBank mRNA BC013081.1 - 3 - 3 352 81
CB155600.1 - 3 - 3 399 81

View Full Product Details