The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002241.4 | NP_002232.2 | 2 - 2 | 4386 | 101 |
GenBank mRNA | BC034036.1 | - | 2 - 2 | 4282 | 101 |
BC037840.2 | - | 1 - 1 | 1396 | 101 | |
BE466069.1 | - | 1 - 1 | 101 |