Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LYSMD4
Entrez Gene ID
145748
Gene Name
LysM domain containing 4
Gene Aliases
-
Chromosome Location
Chr.15: 99715033 - 99733561 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284417.1 NP_001271346.1 3 - 3 2506 120
NM_001284418.1 NP_001271347.1 3 - 3 2507 120
NM_001284419.1 NP_001271348.1 5 - 5 2582 120
NM_001284420.1 NP_001271349.1 2 - 2 2216 120
NM_001284421.1 NP_001271350.1 4 - 4 2532 120
NM_001284422.1 NP_001271351.1 5 - 5 2632 120
NM_152449.3 NP_689662.2 6 - 6 2697 120
XM_011521241.1 XP_011519543.1 2 - 2 2290 120
XM_011521244.1 XP_011519546.1 3 - 3 2859 120
XM_017021926.1 XP_016877415.1 4 - 4 2723 120
XM_017021927.1 XP_016877416.1 3 - 3 3044 120
XM_017021928.1 XP_016877417.1 2 - 2 2688 120
XM_017021929.1 XP_016877418.1 4 - 4 2885 120
XM_017021930.1 XP_016877419.1 4 - 4 2899 120
GenBank mRNA AK057570.1 - 1 - 1 2041 120
AK124852.1 - 4 - 4 2835 120
BC041097.1 - 6 - 6 2674 120
BC084545.1 - 3 - 3 2474 120

View Full Product Details