The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194247.2 | NP_919223.1 | 4 - 5 | 658 | 131 |
XM_005246380.1 | XP_005246437.1 | 4 - 5 | 658 | 131 | |
XM_005246381.4 | XP_005246438.1 | 4 - 5 | 564 | 131 | |
XM_006712365.2 | XP_006712428.1 | 4 - 5 | 658 | 131 | |
XM_017003587.1 | XP_016859076.1 | 4 - 5 | 564 | 131 | |
XM_017003588.1 | XP_016859077.1 | 4 - 5 | 658 | 131 | |
GenBank mRNA | AK293125.1 | - | 4 - 5 | 526 | 131 |
AK304687.1 | - | 4 - 5 | 521 | 131 | |
AK316434.1 | - | 4 - 5 | 526 | 131 | |
AY363225.1 | - | 4 - 5 | 552 | 131 | |
BC064494.1 | - | 4 - 5 | 570 | 131 | |
BC112027.1 | - | 4 - 5 | 580 | 131 | |
BC113470.1 | - | 4 - 5 | 580 | 131 |